Cole-Parmer Ultrasonic Processor Booster
Representative image only.
Cole-Parmer Item # WW-04710-84
For difficult samples
  • Do not use with microtips or high-gain probes
  • Booster for use to process difficult samples
  • 1 year warranty
₹77,500.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Length (in)5
  • Length (cm)12.9
  • DescriptionUltrasonic Processor Booster
  • Warranty1 year
1 Year Warranty
Key Features
  • Do not use with microtips or high-gain probes
  • Booster for use to process difficult samples
  • 1 year warranty
More About this Item
Use this booster to process very difficult samples. It increases the amplitude of vibration at the probe tip by a factor of two.