Cole-Parmer Two-Prong Support Clamp, Single Adjustment, Nickel-Plated Zinc
Representative image only.
Cole-Parmer Item # WW-08027-10
Provide support for your specific laboratory equipment
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Clamp MaterialNickel-plated Zinc
  • Clamp TypeEquipment, Support
  • Length (in)5.99
  • DescriptionTwo-Prong Support Clamp, Single Adjustment, Nickel-Plated Zinc
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Supports your electrolysis apparatus, glass rods, or tubing at any angle. Clamps include integral connector and both slip-on vinyl and fiberglass sleeves.