Cole-Parmer Tuff-Tip In-Line ORP Probe, SJ/PVDF/10'/BNC
Representative image only.
Cole-Parmer Item # WW-05995-20
Semi-domed, tuff-tip limits glass exposure and protects electrode from damage
  • Order single or double junction disposable electrodes for your in-line process control needs
  • Available with BNC or tinned ends
  • 1 year warranty
PRODUCT OPTIONS
₹26,500.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • ApplicationsORP
  • Electrode TypeSingle junction
  • Electrode Body MaterialPVDF
  • Min Temperature (° C)-5
  • Max Temperature (° C)80
  • Thread Size NPT(M) (in)1/2
  • ConnectorBNC
  • Cable Length (m)3
  • DescriptionTuff-Tip In-Line ORP Probe, SJ/PVDF/10'/BNC
  • Warranty1 year
1 Year WarrantyKeep a Spare
Key Features
  • Order single or double junction disposable electrodes for your in-line process control needs
  • Available with BNC or tinned ends
  • 1 year warranty
More About this Item
Electrodes feature PVDF housing with 1/2'' NPT(M) threads and withstands temperatures from 23 to 176°F (--5 to 80°C) and pressures to 100 psi. Electrodes include 10-ft cable with BNC pH connector and tinned ends for ATC element. Insertion depth 1''.