This item is now discontinued. We recommend the following as suggested replacements: WW-35660-16
Traceable Dual-Display Conductivity Meter with Calibration
Representative image only.
Traceable Item # WW-19601-08
Simultaneously displays conductivity and temperature readings
  • Includes NIST-Traceable certificate from our A2LA accredited (1750.01) calibration laboratory
  • Accuracy of ± 2% full-scale plus one digit
  • Automatic and manual temperature compensation
  • Minimum, maximum, and hold functions
  • Serial RS-232 output allow connection to computer or optional datalogger
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Meter TypePortable
  • Display typeLCD
  • DescriptionTraceable® Dual-Display Conductivity Meter with Calibration
  • Warranty1 Year
1 Year WarrantyConformite EuropeenneNIST Traceable Calibration
Key Features
  • Includes NIST-Traceable certificate from our A2LA accredited (1750.01) calibration laboratory
  • Accuracy of ± 2% full-scale plus one digit
  • Automatic and manual temperature compensation
  • Minimum, maximum, and hold functions
  • Serial RS-232 output allow connection to computer or optional datalogger
  • 1 year warranty
More About this Item
Traceable® Products have an individually-numbered Traceable® Certificate provided with each unit, that assures accuracy from our ISO/IEC 17025:2017(1750.01) calibration laboratory accredited by A2LA. It indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMI) that are signatories to the CIPM Mutual Recognition Agreement. Each product goes through an intense vetting process before it can carry the Traceable name, thus saving you both time and money by not having to calibrate separately.