Cole-Parmer Vial Cap Seal for Screw Thread Closure, Clear, Molded 10 mil Septa, 8-425 mm; 100/Pk
Representative image only.
Cole-Parmer Item # WW-98721-10
Keep your samples safe with these vial accessories
  • Select from a variety of caps, septa and inserts that fit your application and needs.
  • This screw thread cap and speta is the closure only—vial sold separately
  • Limited Lifetime Warranty
PRODUCT OPTIONS
₹1,500.00INR / Pkg of 100
SPECS & DESCRIPTION
Specifications & Description
  • Cap TypeScrew-Thread
  • Cap Diameter (mm)8
  • Cap colorClear
  • DescriptionTop Seal Screw Thread Closure, Clear, Molded 10 mil Septa, 8-425 mm
  • WarrantyLimited Lifetime
Limited Lifetime Warranty
Key Features
  • Select from a variety of caps, septa and inserts that fit your application and needs.
  • This screw thread cap and speta is the closure only—vial sold separately
  • Limited Lifetime Warranty
More About this Item
Suitable for use with Cole-Parmer® Versi-Vials
The Top Seal closure with a 10 mil molded septa allows for easy needle penetration and is inert to most solvents. This one piece design saves time by eliminating the process of assembling liners in the closures.