This item is now discontinued. We recommend the following as suggested replacements: WW-24503-04
Cole-Parmer Tips Mini Stacks 960/pk .1-10uL Natural
Representative image only.
Cole-Parmer Item # WW-07911-40
Economical and reduces plastic waste
  • Quickly reloads
  • Universal tips fit most pipettors
  • RNase- and DNase-free
SPECS & DESCRIPTION
Specifications & Description
  • SterileNo
  • Max Volume (µL)10
  • Min Volume (µL)0.1
  • Min Volume (mL)0.001
  • Max Volume (mL)0.01
  • Min Dispensing (mL)0.0001
  • Max Dispensing (mL)0.01
  • Color CodeNatural
  • DescriptionTips Mini Stacks 960/pk .1-10uL Natural
Key Features
  • Quickly reloads
  • Universal tips fit most pipettors
  • RNase- and DNase-free
More About this Item
Ecopette pipette tip reloading system is environmentally friendly and economical by reducing the amount of plastic used to store pipette tips. Quickly refill your pipette tip trays with already pre-racked tips simply by discarding empty tray and inserting cover over new rack. Tips are designed with a universal fit to be used with most pipettors. Available in full stacks of 10 trays or half stacks of 5 trays.