Cole-Parmer TimeStrip Plus, 10°C, 48 Hrs; 50/Pk
Representative image only.
Cole-Parmer Item # WW-15961-49
Timestrip® PLUS indicators are accurate, cost-effective and easy to use
  • Timestrip® PLUS indicators can be directly applied to products and/or packaging
  • These indicators are available for a variety of temperature thresholds ranging from -20°C to +30°C
  • Custom temperatures can be produced subject to minimum order quantities.
PRODUCT OPTIONS
₹8,900.00INR / Pkg
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionTimeStrip Plus, 10°C, 48 Hrs; 50/Pk
Key Features
  • Timestrip® PLUS indicators can be directly applied to products and/or packaging
  • These indicators are available for a variety of temperature thresholds ranging from -20°C to +30°C
  • Custom temperatures can be produced subject to minimum order quantities.