Cole-Parmer Essentials Testing Sieve, Brass Frame/SS Wire, 8" OD, Half Height, No. 30
Representative image only.
Cole-Parmer Essentials Item # WW-59941-08
  • Economical brass frame combined with the durability of stainless steel mesh
  • 1 year warranty
PRODUCT OPTIONS
₹13,700.00INR / Each
₹27,000.00 - ₹33,700.00INR / Each
for an additional
₹27,000.00INR / Each
for an additional
₹33,700.00INR / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • MaterialBrass frame, stainless steel wire
  • U.S. Standard Screen Size (No.)30
  • Tyler Standard Screen Size (mesh)28
  • Sieve Opening (mm)0.595
  • Outside Diameter (in)8
  • Height (InchShort)1
  • DescriptionTesting Sieve, Brass Frame/SS Wire, 8\" OD, half height, No. 30
  • Warranty1 Year
1 Year WarrantyISO9001 Certified Supplier
Key Features
  • Economical brass frame combined with the durability of stainless steel mesh
  • 1 year warranty
More About this Item
These brass and stainless steel (SS) testing sieves are serialized and come with a test sieve certificate of compliance. Equivalent to ASTM E11-09 compliance sieve.