Cole-Parmer Essentials Testing Sieve, 8" SS Frame/SS Wire, Full Height, No. 270
Representative image only.
Cole-Parmer Essentials Item # WW-59984-22
Don't second guess accuracy
  • Come with test sieve certificate of compliance
  • Serialized
  • 1 year warranty
PRODUCT OPTIONS
₹27,600.00INR / Each
₹27,000.00 - ₹33,700.00INR / Each
for an additional
₹27,000.00INR / Each
for an additional
₹33,700.00INR / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • MaterialStainless Steel
  • U.S. Standard Screen Size (No.)270
  • Tyler Standard Screen Size (mesh)270
  • Sieve Opening (mm)0.053
  • Outside Diameter (in)8
  • Height (InchShort)2
  • DescriptionTesting Sieve, 8\" SS Frame/SS Wire, full height, No. 270
  • Warranty1 Year
1 Year WarrantyISO9001 Certified Supplier
Key Features
  • Come with test sieve certificate of compliance
  • Serialized
  • 1 year warranty
More About this Item
These stainless steel (SS) testing sieves are serialized and come with a test sieve certificate of compliance. Equivalent to ASTM E11-09 compliance sieve.