Cole-Parmer Submarine Gel System 2-Place Gel Caster Tray for 28561-00
Representative image only.
Cole-Parmer Item # WW-28561-62
  • 3 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionSubmarine Gel System 2-Place Gel Caster Tray for 28561-00
  • Warranty3 year
3 Year Warranty
Key Features
  • 3 year warranty
More About this Item
Gel Casters allow you to precast consistently flat, even gels using the gel trays sold seperately. Casters accommodate either one or two trays. Casters feature a three-point leveling base and soft, coated neoprene side gaskets that secure trays in place and effectively seal their ends during casting — eliminating the need for taping.