Cole-Parmer Essentials Static-Away Brush, 1-1/2" Handle, 2-1/2" Brush Width
Representative image only.
Cole-Parmer Essentials Item # WW-84551-01
Dissipate static—protects sensitive lab work
  • 1 year warranty
PRODUCT OPTIONS
₹4,000.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • ColorBlack
  • Handle Length (in)1.5
  • Width (in)2 1/2
  • Width (cm)6.35
  • DescriptionStatic-Away Brush, 2-1/2\" Brush, 1-1/2\" Handle; 1/Pk
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Brushes dissipate static while whisking away harmful dust, lint, and dirt from that could damage sensitive lab work. Synthetic fibers won't scratch critical surfaces. Handle is made from a conductive composite material.