Cole-Parmer StableTemp Water Bath Thermometer, Glass; -20 to 150C
Representative image only.
Cole-Parmer StableTemp Item # WW-08128-98
  • 2-1/2 Years Parts and Labor Warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionWater Bath Thermometer, Glass; -20 to 150C
  • Warranty2-1/2 Years Parts and Labor
2 and a Half Year Warranty
Key Features
  • 2-1/2 Years Parts and Labor Warranty
More About this Item
Alcohol thermometer offers a safe alternative to checking you bath temperatures between -20 to +150°C.
For use with:
Circulating Baths
Stainless Steel Open Bath Systems
Polycarbonate Open Bath Systems
Specialty Products
Cryoprecipitate Bath