This item is now discontinued. We recommend the following as suggested replacements: WW-36620-09

Cole-Parmer StableTemp Dry Block Heaters (Digital), 2 Block, 120V

Representative image only.
Cole-Parmer StableTemp Item # WW-36600-03
Precision temperature stability ensures reproducibility of results
  • Create a versatile system unique to your application; order a digital heater and the required heating block module (sold separately online).
  • Interchangeable modular heating blocks provide flexibility for heating different-size tubes -order blocks separately
  • 2 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • Number Of Blocks2
  • Uniformity (° F)±0.3°
  • Uniformity (° C)±0.2°
  • Max Temperature (° F)248
  • Max Temperature (° C)120
  • Power (VAC)120
  • Power (watts)210
  • Height (in)3 1/2
  • Width (in)8
  • Length (in)15 2/5
  • DescriptionDry Block Heaters (Digital), 2 Block, 120V
  • Warranty2 Years
Two year warrantyConformite Europeenne

Key Features

  • Create a versatile system unique to your application; order a digital heater and the required heating block module (sold separately online).
  • Interchangeable modular heating blocks provide flexibility for heating different-size tubes -order blocks separately
  • 2 year warranty

More About this Item

Designed for incubation and activation of cultures, enzyme reactions, immunoassays, melting/boiling points, and a wide variety of other laboratory procedures. Each heater accepts separate interchangeable modular heating blocks, which accommodate various sample enclosures such as microtubes, centrifuge tubes, vials, microplates, and PCR strips or tubes. Modular blocks are constructed from a solid anodized aluminum block. The close tube to block walls allow for maximum heat retention. Each block has a thermometer well for measuring block temperature. Optional PT100 RTD temperature probe kit enables the unit to read the actual block or sample temperature and display the temperature on the control panel. Create a versatile system unique to your application; order either an analog or digital heater and the required heating block module (sold separately online).

CAUTION! To avoid possible electrical hazard, do not fill well or block opening with water or other fluids. Unit is designed to run as a dry bath / incubator.

Digital Dry Block Heaters offer exceptional temperature uniformity and feature a proportional integrative derivative (PID) controller. PID control allows you to select a particular temperature with ease (in 0.1°C increments), enables rapid heat-up for samples, and provides excellent stability to maintain results. Features include timer, audible alarm, built-in temperature calibration mode, overshoot protection, and independent LED displays that show actual and setpoint temperatures

.Digital dry block heater 2 block capacity 120V with independent LED displays for temperature & time.PID temperature control with user calibration mode.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat