Cole-Parmer StableTemp Digital Water Bath Flask Holder; 25 mL
Representative image only.
Cole-Parmer StableTemp Item # WW-12504-91
Optimize performance of your StableTemp Digital Utility Water Bath
  • Keep your running properly with these flask holders and platforms
  • 2-1/2 Years Parts and Labor Warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionDigital Water Bath Flask Holder; 25 mL
  • Warranty2-1/2 Years Parts and Labor
2 and a Half Year Warranty
Key Features
  • Keep your running properly with these flask holders and platforms
  • 2-1/2 Years Parts and Labor Warranty
More About this Item
Choose from a wide range of replacement parts and accessories to ensure top performance of your StableTemp Digital Utility Water Bath.
Compatible with the 20 and 28 liter shaking water baths, these clips screw into the (included) perforated platform. Clips are sold individually; one clip holds one flask.