Cole-Parmer Essentials Spatula, "Reagent Digger", Stainless Steel, 6.5" L; 12/PK

Representative image only.
Cole-Parmer Essentials Item # WW-01019-13
Shaped to fit reagent vessels
  • Pointed end easily handles small amounts
  • Length 6 1/2" (16.5 cm)

₹5,500.00INR / Pkg of 12

SPECS & DESCRIPTION

Specifications & Description

  • Overall Length (in)6 1/2
  • Overall Length (cm)16.5
  • MaterialStainless Steel
  • SterileNo
  • DescriptionSpatula, \"Reagent Digger\", 304SS, 6.5\" L, 12/pk

Key Features

  • Pointed end easily handles small amounts
  • Length 6 1/2" (16.5 cm)

More About this Item

Facilitates scooping and measuring of powdered, flaked, crystalline, or granular media—especially useful for dislodging caked or crystallized reagents. Uniquely shaped stainless steel spatula fits reagent vessels with openings as small as 15⁄16" (2.4 cm) dia.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat