Cole-Parmer Vial Cap, Snap Top, Clear, PTFE/Red Butyl Rubber Liner, 11 mm; 100/Pk
Representative image only.
Cole-Parmer Item # WW-98720-16
Keep your samples safe with these vial accessories
  • Select from a variety of caps, septa and inserts that fit your application and needs.
  • This snap top cap and liner is the closure only—vial sold separately
  • Limited Lifetime Warranty
PRODUCT OPTIONS
₹4,100.00INR / Pkg of 100
SPECS & DESCRIPTION
Specifications & Description
  • Cap TypeSnap Top
  • Cap Diameter (mm)11
  • Cap colorClear
  • DescriptionSnap Top Cap, Clear PTFE/Red Butyl Rubber Lined, 11 mm
  • WarrantyLimited Lifetime
Limited Lifetime Warranty
Key Features
  • Select from a variety of caps, septa and inserts that fit your application and needs.
  • This snap top cap and liner is the closure only—vial sold separately
  • Limited Lifetime Warranty
More About this Item
Suitable for use with Cole-Parmer® Versi-Vials
Snap Top Caps can be snapped on and off by hand which eliminates the need for metal crimping and decapping tools, reducing potential hazards due to sharp aluminum edges. Large opening in top provides greater target area for needle penetration.