Cole-Parmer Single-Stage Low-Pressure Specialty Regulator; CGA 660
Representative image only.
Cole-Parmer Item # WW-03274-10
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Max Delivery Pressure (psig)50
  • Max Inlet Pressure (PSI)400
  • Outlet Connections¼\" NPT(M)
  • CGA Fitting660
  • Capacity (SCFH)2200
  • Outlet Gauge Graduations (PSI)2
  • Inlet Gauge Graduations (PSI)100
  • DescriptionSingle-Stage Low-Pressure Specialty Regulator; CGA 660
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Model 03274-10 is a single-stage unit specifically designed for gases that attack brass or copper-bearing alloys. When used with low-vapor-pressure liquefied gases, no inlet gauge is needed. Rubber-supported PTFE diaphragm, KEL-F seat, aluminum-alloy body and bonnet, SS delivery gauge, and 10 mm sintered SS filter.