Cole-Parmer Essentials Sieve; No 270, Half-Height, 8", SS/SS, NIST-Traceable Calibration Certificate
Representative image only.
Cole-Parmer Essentials Item # WW-59985-72
Sieve openings are measured with a computerized video imaging system
  • 1 year warranty
PRODUCT OPTIONS
₹47,500.00INR / Each
₹27,000.00 - ₹33,700.00INR / Each
for an additional
₹27,000.00INR / Each
for an additional
₹33,700.00INR / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • MaterialStainless Steel
  • U.S. Standard Screen Size (No.)270
  • Tyler Standard Screen Size (mesh)270
  • Sieve Opening (mm)0.053
  • Outside Diameter (in)8
  • Height (InchShort)1
  • DescriptionSieve; No 270, Half-Height, 8\", SS/SS, NIST Cal Cert
  • Warranty1 Year
1 Year WarrantyNIST Traceable Calibration
Key Features
  • 1 year warranty
More About this Item
Sieve is serialized, comes with a test sieve certificate, and meets ASTM E11 specifications. Includes an NIST-traceable calibration report supplied by the manufacturer.