Cole-Parmer Vial Cap Seal for Screw Thread Closure, PTFE/Red Rubber, 8-425 mm; 100/Pk
Representative image only.
Cole-Parmer Item # WW-98721-21
Keep your samples safe with these vial accessories
  • Select from a variety of caps, septa and inserts that fit your application and needs.
  • This seal is compatible with cap 98721-12 sold separately
  • Seal has 0.035" thickness
  • Limited Lifetime Warranty
PRODUCT OPTIONS
₹2,300.00INR / Pkg of 100
SPECS & DESCRIPTION
Specifications & Description
  • Cap TypeScrew-Thread
  • Cap Diameter (mm)8
  • Cap colorRed
  • DescriptionSeals for Screw Thread Closure, PTFE/Red Rubber, 8-425 mm
  • WarrantyLimited Lifetime
Limited Lifetime Warranty
Key Features
  • Select from a variety of caps, septa and inserts that fit your application and needs.
  • This seal is compatible with cap 98721-12 sold separately
  • Seal has 0.035" thickness
  • Limited Lifetime Warranty
More About this Item
Suitable for use with Cole-Parmer® Versi-Vials
PTFE/Red rubber Septa for use with Screw Top Closure #98721-12.