Cole-Parmer Sample Stirring Kit; HDPE
Representative image only.
Cole-Parmer Item # WW-66501-20
Versatile sampling kits with interchangeable parts
  • All components made of FDA-compliant HDPE
  • Purchase individual replacement components as needed!
SPECS & DESCRIPTION
Specifications & Description
  • MaterialHDPE
  • SterileNo
  • DescriptionSample Stirring Kit; HDPE
Key Features
  • All components made of FDA-compliant HDPE
  • Purchase individual replacement components as needed!
More About this Item
All dipping beakers, stir blades, and sampling bottles use the same thread to be compatible with all three handle length options. Replacement handles and samplers may be replaced as needed — no need to purchase an entire kit if a part is lost or worn!
Sample Stirring Kit includes:
One 1.5 ft (0.45 m) handle
One 3 ft (0.9 m) handle
One 6 ft (1.8 m) handle
One solid stirring blade
One perforated stirring blade
The stirring blades are threaded to connect to any of the handles. Use the solid stirring blade for more viscous samples, and the perforated blade for mixing thinner liquids.