Cole-Parmer Essentials RR-700 Monobed Deionizer Mix-Bed Resin, Nuclear-Grade with Color Indicator
Representative image only.
Cole-Parmer Essentials Item # WW-01503-30
Keep your monbed deionizer running smoothly
  • Designed to be easily interchanged with your monobed deionizer
  • 1 year warranty
PRODUCT OPTIONS
₹21,900.00INR / Each
Often Bought with…
SPECS & DESCRIPTION
Specifications & Description
  • Compatible WithMonobed Deionizer System
  • DescriptionResin, nuclear-grade with color indicator
  • Warranty1 year
1 Year Warranty
Key Features
  • Designed to be easily interchanged with your monobed deionizer
  • 1 year warranty
More About this Item
Choose from resins and an ultrapure cartidge to provide high purity water from your monobed deionizer water systems.
The mixed-bed resin is made up of a cation and anion bead allowing water to flow through the deionizer tanks and come out near zero totatl dissolved solids, creating deionized water.
The resin is exhausted when chlorine/chlorides leach out. To test, take a beaker of water from the demineralizer and add a drop of silver chloride. If the water is cloudy then the resin should be changed.