Cole-Parmer Replacement Fan Filter
Representative image only.
Cole-Parmer Item # WW-55008-14
Replacement parts keep your overhead mixer running smoothly
PRODUCT OPTIONS
₹11,600.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionReplacement Fan Filter
More About this Item
The fan filter is removeable, washable for cleaning, and replaceable. When cleaning the fan filter, proceed as appropriate based on materials in contact with the filter. Avoid use of harsh scrubbers or brushes that could damage the mesh. Ensure the mesh is intact and dry before installing into the SD controller.