Cole-Parmer Rack insert for BOD bottles; inserts on either top or bottom rack, half size
Representative image only.
Cole-Parmer Item # WW-98951-60
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Compatible With98950-01, 98950-11, 98950-20, 98950-30
  • DescriptionRack insert for BOD bottles; inserts on either top or bottom rack, half size
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Insert fits on either the upper or lower general purpose racks included with the Cole-Parmer Glassware Washers. Half size insert will take up half of a rack, and holds up to 18 BOD bottles.