This item is now discontinued. We recommend the following as suggested replacements: WW-98075-36
Cole-Parmer Pressure Switch, 5 To 25 PSI, Normally Open Spst; 1/4" NPT(M) Brass Fitting
Representative image only.
Cole-Parmer Item # WW-68973-50
Low cost and short lead times make this switch perfect for OEM applications
  • Ideal for pneumatic and low pressure hydraulic applications
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Min Pressure (PSI)5
  • Max Pressure (PSI)25
  • Wetted MaterialsBrass, Kapton®, nitrile
  • Process Connection1/4\" NPT(M)
  • Battery Type100 VDC
  • Electrical Connectionsexposed terminals
  • Switch outputN.O.
  • OutputSPST
  • Media CompatibilityGases and liquids compatible with wetted materials
  • DescriptionPressure Switch, 5 To 25 PSI, Normally Open Spst; 1/4\" NPT(M) Brass Fitting
  • Warranty1 year
1 Year WarrantyConformite Europeenne
Key Features
  • Ideal for pneumatic and low pressure hydraulic applications
  • 1 year warranty
More About this Item
These compact pressure switches are designed for OEM applications. Made economical with metal blade contacts instead of microswitches, these switches feature Kapton® diaphragms which maintain excellent physical properties over a wide temperature range, as well as offer superb chemical resistance with organic solvents.