Cole-Parmer Preconfigured Modular Submersible pH Kit
Representative image only.
Cole-Parmer Item # WW-27113-04
Preconfigured pH kits make it easy to save; modular design reuses housing and replaces electrode as needed
  • Flat-surface electrode design minimizes electrode damage
  • Self-cleaning feature extends life of electrode
  • Choose from submersible and in-line kits
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • ApplicationsSelf-cleaning
  • Electrode TypeDouble junction
  • Electrode Body MaterialRyton® PPS
  • Reference JunctionCeramic
  • Temperature CompensationYes
  • ATC ConnectorTinned ends
  • ConnectorBNC
  • DescriptionPreconfigured Modular Submersible pH Kit
  • Warranty1 year
1 Year Warranty
Key Features
  • Flat-surface electrode design minimizes electrode damage
  • Self-cleaning feature extends life of electrode
  • Choose from submersible and in-line kits
  • 1 year warranty
More About this Item
Purchase a pre-configured kits to meet your application requirements. Economical alternative replaces cartridge and reuses housing and cables.
These self-cleaning, flat surface electrodes have been enhanced with protection against thermal and chemical breakdown. The double-junction pH reference protects from heavy metals, sulfur, iodine, or proteins.
Kit for general purpose applications includes cap and cable (27113-00), adapter (27113-20), and pH electrode (27113-30).