Cole-Parmer Essentials Precision Stainless Steel Tweezers, Strong Thick Tips, OO-SA, 120 mm
Representative image only.
Cole-Parmer Essentials Item # WW-07387-00
Ideal for precision handling applications
  • Antimagnetic and antiacid stainless steel construction
  • Rugged and corrosion-resistant design
PRODUCT OPTIONS
₹1,900.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Overall Length (in)4 3/4
  • Overall Length (cm)12
  • MaterialStainless Steel Antimagnetic
  • SterileNo
  • Style0 Strong Straight Thick, Flat Tips
  • DescriptionPrecision Stainless Steel Tweezers, Strong Thick Tips, OO-SA, 120 mm
Key Features
  • Antimagnetic and antiacid stainless steel construction
  • Rugged and corrosion-resistant design
More About this Item
These precision stainless steel tweezers offer good corrosion resistance to most chemicals, salts, and acids. Great for lab work such as handling samples or specimens, and observing them under a microscope. The antimagnetic nature of the material makes these tweezers useful in the electronic, watch, and jewelry industries.