Cole-Parmer Plexiglas Racks for 30 Tubes with 13 mm diameter
Representative image only.
Cole-Parmer Item # WW-01137-12
Interchangeable racks hold a variety of tubes and vials
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionPlexiglas Racks for 30 Tubes with 13 mm diameter
More About this Item
Plexiglas Racks for 6- and 8-L Tanks are available with different hole diameters to hold test tubes, scintillation vials, or 1.5-mL microcentrifuge tubes. Racks attach to the tank sides and suspend tubes or vials in bath. The 6-L tank holds up to three racks without a circulator or two racks with a circulator. The 8-L tank holds up to four racks without a circulator or three racks with a circulator. Circulator sold separately.