Cole-Parmer Pipette Tips 50 to 1000 µl; PP, clear, graduated, sterile, 10 refill racks, 960/pk
Representative image only.
Cole-Parmer Item # WW-25712-58
Cleanroom produced for ready to use applications
  • Constructed of pure, high-quality polypropylene without additives
  • Certified RNase, DNase, and endotoxin free
  • Autoclavable at 121°C for 20 minutes
SPECS & DESCRIPTION
Specifications & Description
  • SterileYes
  • Max Volume (µL)1000
  • Min Volume (µL)50
  • Overall Length (in)2.75
  • Overall Length (mm)70
  • Wetted MatterialPolypropylene
  • Package Of10 refill racks of 96 each
  • DescriptionPipette Tips 50 to 1000 µl; PP, clear, graduated, sterile, 10 refill racks, 960/pk
Key Features
  • Constructed of pure, high-quality polypropylene without additives
  • Certified RNase, DNase, and endotoxin free
  • Autoclavable at 121°C for 20 minutes
More About this Item
The tips are available in non-sterile and sterile rack options, as well as non-sterile and sterile stacked rack options. These are designed to give high levels of accuracy, precision, and reproducibility for challenging applications. The smooth surface of the tips minimizes binding of enzymes and nucleic acids to the walls. All tips are suitable for use with common market pipettes.