Cole-Parmer Pipette Filter Tips 50 to 1000 µl; PP, clear, graduated, sterile, 10 refill racks, 960/pk
Representative image only.
Cole-Parmer Item # WW-25712-92
Reduce waste by refilling
  • Cleanroom produced for ready-to-use applications
  • Slim, thin-walled tips feature graduations
  • Filter protects samples and pipettor against contamination
  • Certified RNase, DNase, and endotoxin free
  • Autoclavable at 121°C for 20 minutes
SPECS & DESCRIPTION
Specifications & Description
  • SterileYes
  • Max Volume (µL)1000
  • AutoclavableYes
  • Min Volume (µL)50
  • Overall Length (in)3.25
  • Overall Length (mm)82
  • Body MaterialPP
  • Color CodeClear
  • Filter TypeNon-self-sealing
  • Package Of10 racks of 96 each
  • DescriptionPipette Filter Tips 50 to 1000 µl; PP, clear, graduated, sterile, 10 replacement racks, 960/pk
Sterile
Key Features
  • Cleanroom produced for ready-to-use applications
  • Slim, thin-walled tips feature graduations
  • Filter protects samples and pipettor against contamination
  • Certified RNase, DNase, and endotoxin free
  • Autoclavable at 121°C for 20 minutes
More About this Item
Constructed of pure, high-quality polypropylene (PP) without additives, these tips are designed to give high levels of accuracy, precision, and reproducibility. The smooth surface minimizes binding of enzymes and nucleic acids to the tip wall.
The non-self-sealing filter plug prevents aerosols and liquids from being aspirated into the body of the pipettor. Filter is free from chemical additives, does not swell, and is made of pure polyethylene (PE).
Each 96-tip refill rack comes in an environmentally friendly package—simply insert rack into an existing storage box.