Cole-Parmer pHASE Combination Double Junction Sealed pH Electrode, BNC connector
Representative image only.
Cole-Parmer Item # WW-27102-91
  • 30" cable length
  • 1 year warranty
₹56,000.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • ApplicationsSealed
  • Electrode TypeDouble junction
  • Electrode Body MaterialEpoxy
  • Min pH (pH)14
  • Max Temperature (° C)100
  • ConnectorBNC
  • DescriptionpHASE Combination Double Junction Sealed pH Electrode, BNC connector
  • Warranty1 year
1 Year Warranty
Key Features
  • 30" cable length
  • 1 year warranty
More About this Item
Epoxy bodies provide maximum ruggedness and good chemical resistance. pHASE electrodes are supplied standard with a 30 inch (76 cm) cable and BNC connector.