Cole-Parmer PH5 pH Pocket Tester
Representative image only.
Cole-Parmer Item # WW-15961-61
Low-cost pocket tester perfect for less demanding applications
  • Economical and easy to use
  • Replaceable single-junction sensor saves money
  • IP67- rated floatable meter – accidental drops into water will not result in lost or damaged meters
  • Push-button calibration with auto-buffer recognition
  • Waterproof, dustproof housing designed for use in dirty and damp field conditions
SPECS & DESCRIPTION
Specifications & Description
Key Features
  • Economical and easy to use
  • Replaceable single-junction sensor saves money
  • IP67- rated floatable meter – accidental drops into water will not result in lost or damaged meters
  • Push-button calibration with auto-buffer recognition
  • Waterproof, dustproof housing designed for use in dirty and damp field conditions
More About this Item
Premium Line Pocket Tester -pH5 Pocket features a replaceable single junction sensor with a recessed bulb to protect against breakage. The unit also offers up to three-point calibration with auto-buffer recognition which allows calibration at 3 different points (pH 4, 7 and 10) to give you ±0.1 pH accuracy for value model and ±0.01 pH for premium model pH1 and across the acid and alkali ranges, self-diagnostic error messages, stable reading indication. Tester automatically switches off in 8 minutes upon no operation.