Cole-Parmer pH flow-through cell, 6000 uL, 7 mm ID, glass
Representative image only.
Cole-Parmer Item # WW-05662-40
Ideal for fermentation applications
  • Accurate to ±0.1 pH units
  • Choose 316 stainless steel cell for long-lasting durability
  • 1 year warranty
PRODUCT OPTIONS
₹20,800.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Electrode Body MaterialGlass
  • DescriptionpH flow-through cell, 6000 uL, 7 mm ID, glass
  • Warranty1 year
1 Year Warranty
Key Features
  • Accurate to ±0.1 pH units
  • Choose 316 stainless steel cell for long-lasting durability
  • 1 year warranty
More About this Item
Measure the pH of high-purity water without the problems usually associated with low-ionic strength solutions.
Not a complete system (electrode and cable needed). See accessories section for required extra components.