Cole-Parmer pH electrode refillable single-junction for Beckman meters
Representative image only.
Cole-Parmer Item # WW-27507-08
Save money without giving up temperature compensation!
  • Electrodes designed to work with meters you already have
  • Seven different designs available—find the design that works best for your application without losing automatic temperature compensation
  • 1 year warranty
PRODUCT OPTIONS
₹18,100.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • ApplicationsRefillable
  • Electrode TypeSingle junction
  • Electrode Body MaterialGlass
  • ConnectorBNC
  • DescriptionpH electrode refillable single-junction for Beckman meters
  • Warranty1 year
1 Year Warranty
Key Features
  • Electrodes designed to work with meters you already have
  • Seven different designs available—find the design that works best for your application without losing automatic temperature compensation
  • 1 year warranty
More About this Item
“All-in-One” electrodes offer versatility and customization. These electrodes combine the pH reference, pH half-cell, and temperature sensor in one 12mm-diameter electrode body. The seven designs allow you to select between low-maintenance, sealed electrodes or clog-resistant refillable electrodes. Tuff-Tip electrodes feature a semi-domed tip which protects the pH bulb and ensures a quick response in harsh and dirty samples. PBR-style electrodes provide predictable, reliable, and quick responses—no matter what the sample temperature. This design virtually eliminates drift, accuracy problems, and premature performance losses due to temperature. All electrodes include 3-ft cable and BNC connector for pH, except where noted.Beckman Instruments-compatible electrodes feature a 30K temperature compensator with a 3.5mm audio plug for connecting to your meter.