Cole-Parmer pH Electrode, Fermentation, 12 x 225 mm, Wick Junction, ATC, 4-pin connection
Representative image only.
Cole-Parmer Item # WW-29045-04
New glass formulation provides unparalled performance!
  • Lightening fast response time with superior accuracy and repeatabilty
  • Perfect for low-ionic strength solutions
  • Provides confident values over the entire pH range from 0 to 14
  • Choose wick junction for nonaqueous, low ionic strength, and strong oxidizers or reducing agent samples
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Electrode TypeDouble junction
  • Electrode Body MaterialGlass
  • Reference JunctionWick
  • Min pH (pH)0
  • Max pH (pH)14
  • Accuracy±0.02 pH
  • Max Temperature (° C)130
  • DescriptionpH Electrode, Fermentation, 12 x 225 mm, Wick Junction, ATC, 4-pin connection
  • Warranty1 year
1 Year Warranty
Key Features
  • Lightening fast response time with superior accuracy and repeatabilty
  • Perfect for low-ionic strength solutions
  • Provides confident values over the entire pH range from 0 to 14
  • Choose wick junction for nonaqueous, low ionic strength, and strong oxidizers or reducing agent samples
  • 1 year warranty
More About this Item
Cole-Parmer's fast-response fermentation pH electrodes are made of a proprietary new formulation of low-impedance glass replaces all other formulas — allows you to streamlined inventories and eliminate wasted time trying to select the best electrode for your application. Use in low-ionic strength solutions, high pH testings where sodium error could be a problem, and at high temperatures up to 266°F (130°C). pH is registered in less than 20 seconds, with a typically accuracy of 0.02 pH units (meter dependant).
Chemical-resistant Ryton ''PPS'' PG13.5 threaded pressure fitting, PTFE spacer, and silicone o-ring ensure a leakproof connection to your assembly.
All electrodes feature double-junction reference for maximum lifespan.
Order detachable cap