Cole-Parmer pH electrode, combination, double-junction, sealed
Representative image only.
Cole-Parmer Item # WW-05662-80
Ideal for fermentation applications
  • Accurate to ±0.1 pH units
  • Choose 316 stainless steel cell for long-lasting durability
  • 1 year warranty
PRODUCT OPTIONS
₹46,900.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Electrode TypeDouble junction
  • Electrode Body MaterialGlass
  • Reference ElectrolyteAg/AgCl
  • ConnectorDepends upon cable purchased separately: BNC or Ingold K9
  • DescriptionpH electrode, combination, double-junction, sealed
  • Warranty1 year
1 Year Warranty
Key Features
  • Accurate to ±0.1 pH units
  • Choose 316 stainless steel cell for long-lasting durability
  • 1 year warranty
More About this Item
Replacement electrodes for your flow-through cell. Select the electrode that is compatible with your model of cell, with sealed and refillable options available.