Cole-Parmer Adhesive Microplate Sealing Film, Nonsterile, Polypropylene; 100/PK
Representative image only.
Cole-Parmer Item # WW-67103-55
Provides exceptional seal across entire plate to safeguard samples during transport or storage
  • Prevents sample evaporation and contamination
  • RNase and DNase free
  • Nonsterile
  • 3 month warranty
₹11,000.00INR / Pkg of 100
SPECS & DESCRIPTION
Specifications & Description
  • Max Temperature (° F)215.6
  • Min Temperature (° F)-40
  • Max Temperature (° C)102
  • Min Temperature (° C)-40
  • DescriptionAdhesive Microplate Sealing Film, Nonsterile, Polypropylene; 100/PK
  • Warranty3 month
3 Month Warranty
Key Features
  • Prevents sample evaporation and contamination
  • RNase and DNase free
  • Nonsterile
  • 3 month warranty
More About this Item
Adhesive microplate sealing film is specifically designed for PCR applications and the sealing of microplates for transport and storage. Constructed of 60 µm thick clear polypropylene film, these microplate sealing films seal tightly to prevent well-to-well contamination and sample evaporation. Suitable for many applications, they can be used from -40 to 102 °C (-40 to 215 °F). Easy to apply; just peel and stick. RNase and DNase free.