PCRmax 96-well Polypropylene PCR Microplate, Unskirted, Clear, Nonsterile; 50/Pk
Representative image only.
PCRmax Mfr # 321-21-052 – Item # WW-67103-65
Designed to amplify nucleic acids during the PCR process
  • Manufactured with 99.9% pure virgin polypropylene—withstands temperatures of –20 to 122°>C and are autoclavable
  • Ultra-thin walls accelerate heat transfer providing shorter cycle times
  • Full-length, even insertion cap ensures a tight fit, eliminating sample evaporation
  • Clear, thin wall well bottom
  • Ultra-thin and consistent wall thickness allows precise thermal transfer for optimal results
  • RNase-/DNase-free, nonpyrogenic, nonsterile
  • Can be sealed with sealing film
PRODUCT OPTIONS
₹35,100.00INR / Pkg of 50
Often Bought with…
SPECS & DESCRIPTION
Specifications & Description
  • Number of Wells96
  • Well Volume (mL)0.2
  • Well Volume (µL)200
  • Well ShapeRound
  • SterileNo
  • Plate TypeUnskirted
  • Plate ColorClear
  • MaterialPolypropylene
  • DescriptionPCR Plates, 96-well, unskirted; 50/pk
Key Features
  • Manufactured with 99.9% pure virgin polypropylene—withstands temperatures of –20 to 122°>C and are autoclavable
  • Ultra-thin walls accelerate heat transfer providing shorter cycle times
  • Full-length, even insertion cap ensures a tight fit, eliminating sample evaporation
  • Clear, thin wall well bottom
  • Ultra-thin and consistent wall thickness allows precise thermal transfer for optimal results
  • RNase-/DNase-free, nonpyrogenic, nonsterile
  • Can be sealed with sealing film
More About this Item
PCRmax® PCR products are specifically designed for amplifying nucleic acids during the Polymerase Chain Reaction (PCR) process. Certified RNase, DNase, and pyrogen safe to ensure no degradation of the target or amplified product.