This item is now discontinued. We recommend the following as suggested replacements: WW-24501-48
Cole-Parmer Omega® Pipette Tip, 0.1 to 10 µL, Reloading Stack, 10 Racks; 960/PK
Representative image only.
Cole-Parmer Item # WW-24501-54
Tips free of human touch is purity at its best
  • Tip Options: Standard or filtered
  • Specially developed PE filters free of chemical additives stops aerosols and contaminations
  • Tips also leach-free, ensuring maximum recovery of sample yield
  • Volume measurements, along the tube are marked in increments for easy identification
  • Produced from Swiss-made Diamond Finish Mold
  • Made of FDA approved special medical grade virgin polypropylene plastic with high transparency
  • Unique label coding in packaging for easy identification
  • Nonsterile, nonfilter tips Volume: 0.1 to 10 µL
SPECS & DESCRIPTION
Specifications & Description
  • SterileNo
  • Max Volume (µL)10
  • Min Volume (µL)0.1
  • Tip StyleStandard, 3.13 cm L
  • Body MaterialPolypropylene (PP)
  • Filter TypeNone
  • Package OfReloading Stack, 10 x 96
  • DescriptionPipette Tip, 0.1 to 10 µL, Reloading Stack, 10 Racks; 960/PK
Bio DNase-FreeNonpyrogenicRNase-Free
Key Features
  • Tip Options: Standard or filtered
  • Specially developed PE filters free of chemical additives stops aerosols and contaminations
  • Tips also leach-free, ensuring maximum recovery of sample yield
  • Volume measurements, along the tube are marked in increments for easy identification
  • Produced from Swiss-made Diamond Finish Mold
  • Made of FDA approved special medical grade virgin polypropylene plastic with high transparency
  • Unique label coding in packaging for easy identification
  • Nonsterile, nonfilter tips Volume: 0.1 to 10 µL
More About this Item
Not your everyday graduated pipette tips. It starts with a touch-free manufacturing facility and stringent quality control process to make these clear, plastic tips certified free of any affecting biological elements, such as human DNA, DNase and RNase and Endotoxin, or Pyrogen and PCR inhibitors. They also are noncytotoxic.
It’s about their innovative design, too. The profile and dimensions of the tips are engineered to accurately measure and transfer a volume of liquid from one container to another, as well as adapt to a wide-range of pipettes.
It ends with preserving their purity in the following packaging options, which are free of lubricants and heavy metals.
Reload Stack for Filter Tips: Supplied in a sterile, sealed and eco-friendly package free of DNase, RNase, and Pyrogen. Several options for the number of tips in each pack are offered.
Individually Wrapped Tips: Free of DNase, RNase and Pyrogen