Cole-Parmer Mixer Assembly with 80 mm cap, 1200 rpm motor, and controller
Representative image only.
Cole-Parmer Item # WW-06064-79
Uniform and homogenous mixing with no dead spots
  • Provides both vertical and horizontal mixing to produce a full mix
  • Elimination of external blades reduces risk of injury during cleaning
  • Full mixing can be achieved within 6 seconds
  • Stainless steel parts can be autoclaved for sterilizing prior to mixing
  • Minimal surface turbulence results in higher energy efficiency
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPP
  • Width (InchShort)5
  • Height (in)6 11/16
  • Max Speed (rpm)1200
  • DescriptionMixer Assembly with 80 mm cap, 1200 rpm motor, and controller
ISO9001 Certified Supplier
Key Features
  • Provides both vertical and horizontal mixing to produce a full mix
  • Elimination of external blades reduces risk of injury during cleaning
  • Full mixing can be achieved within 6 seconds
  • Stainless steel parts can be autoclaved for sterilizing prior to mixing
  • Minimal surface turbulence results in higher energy efficiency
More About this Item
The turbine mixer assembly mixes materials quickly and evenly without dead spots inside carboys. The assembly features a polypropylene cap, 1200 rpm motor, and speed controller. Match different mixing heads and shafts with the mixer assembly to create one of four mixing styles—top anaerobic, top aerobic, full mixing, and bottom mixing. Each mixing style produces an even mix without the formation of a layer of material left behind on the carboy’s wall, or sediment at the bottom of the carboy.
Top anaerobic mixing is for mixing less viscous materials from the top, getting a long fluid motion. Top aerobic mixing is for adding air into the mixture. Full mixing is for simultaneously circulating fluid from top to center and bottom to center. Bottom mixing is for lifting heavy or viscous material siting on the bottom. Autoclavable stainless steel mixing heads are available in 15-, 25-, 50-, and 100-mm sizes.
To create a complete mixing system that meets your needs, order a cap mixer assembly, shaft, mixing head, and connector.