Cole-Parmer Miniature Air Filter and Regulator, 2 to 125 psi, 20 scfm
Representative image only.
Cole-Parmer Item # WW-98252-00
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Min Operating Temperature (° F)40
  • Max Operating Temperature (° F)125
  • Min Operating Temperature (° C)4
  • Max Operating Temperature (° C)52
  • Wetted Materials20 µm porous bronze
  • Outlet Gauge Range (PSI)2 to 125
  • DescriptionMiniature Air Filter and Regulator, 2 to 125 psi, 20 scfm
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Combines filter and regulator in one compact unit. Precision diaphragm gives you high accuracy regulation from 2 to 125 psig (150 psig max). Rated for 20 scfm; operates from 40 to 125ºF (4 to 52ºC). Detachable, 20 µm porous bronze filter provides 98% moisture removal. Overnight drain requires minimal maintenance—automatically releases accumulated condensate when pressure falls below 3 psig. Filter/regulator has 1/4" NPT(F) ports and 1/8" NPT(F) gauge connections. Use with air.