Cole-Parmer Mini Fast Sub, Horizontal Mini Gel Kit

Representative image only.
Cole-Parmer Item # WW-28559-20
Fast, compact mini horizontal system has results in less than an hour!
  • Maximum of 50 mL of agarose necessarysave on reagents
  • UV transparent base allows easy sample visualization
  • 3 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • Buffer Solution Required (mL)50
  • Gel Length (cm)10
  • Gel Width (cm)7.5
  • Cell Width (in)5.5
  • Cell Depth (in)6.6875
  • Cell Width (cm)13.97
  • Cell Height (cm)7.94
  • Cell Depth (cm)16.99
  • DescriptionMini Fast Sub, Horizontal Mini Gel Kit
  • Warranty3 Years
3 Year WarrantyConformite Europeenne

Key Features

  • Maximum of 50 mL of agarose necessarysave on reagents
  • UV transparent base allows easy sample visualization
  • 3 year warranty

More About this Item

The mini-quick horizontal system is designed for easy, simple, and safe runs. The PVC dams can be snapped into the grooves for easy gel casting. Prep work is minimal since gels can be cast in place. The uniplanar design has a buffer chamber only slightly longer than the gel itself, minimizing the amount of buffer required. Use one or two comb positions for up to a max of 40 sample output. The system is ideal for electroelution of DNA in dialysis bags, or separation and identification of DNA fragments, RNA fragments, PCR products, and synthetic oligonucleotides. Less than fifteen minutes of run time is required for elution of DNA in dialysis bags.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat