Cole-Parmer Midi-horizontal gel units, 14 x 10 cm gel bed

Representative image only.
Cole-Parmer Item # WW-28560-00
Reduced gel path length for rapid sequencing of samples
  • 3 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • Buffer Solution Required (mL)700
  • Gel Length (cm)10
  • Gel Width (cm)14
  • Cell Width (in)6.5
  • Cell Height (in)5.5
  • Cell Depth (in)8.5
  • Cell Width (cm)2.56
  • Cell Height (cm)13.97
  • Cell Depth (cm)3.35
  • DescriptionMidi-horizontal gel units, 14 x 10 cm gel bed
  • Warranty3 Years
3 Year Warranty

Key Features

  • 3 year warranty

More About this Item

Tapered-baffle design eliminates the need for sealing or taping. UV-transparent acrylic gel tray has ''anchors'' to prevent gel from floating during electrophoresis. Optional gel casters let you pour gels outside of the unit, and to pour two gels simultaneously.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat