Cole-Parmer Microscope Slide Holder Cabinet; 1 Drawer, 400 Slide Capacity
Representative image only.
Cole-Parmer Item # WW-48580-21
Store microscope slides with ease, speed, and efficiency
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionMicroscope Slide Holder Cabinet; 1 Drawer, 400 Slide Capacity
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Organize and classify your slides for instant reference and fast retrieval. The durable slide holder is 3'' x 5'' cardboard fitted with a protective plastic frame that holds the slides while keeping them complete separated. Each holder accepts four 75 mm x 25 mm or 3'' x 1'' slides of any thickness. Four-position tabs adapt to your slide identification system while four grid lines allow you to add any comments for each slide in the holder.
Optional slide holder cabinets (sold separately) are an easy way to organize your slide holders.
Model 48580-21 accepts 100 slide holders (400 slides total).