Cole-Parmer Micro Spatula Set, 316 SS; 4/Pk
Representative image only.
Cole-Parmer Item # WW-06369-40
  • Ideal for handling extremely small samples
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Overall Length (in)2 1/2
  • Overall Length (cm)6.35
  • MaterialStainless Steel
  • SterileNo
  • DescriptionMicro Spatula Set, 316 SS; 4/Pk
  • Warranty1 year
1 Year Warranty
Key Features
  • Ideal for handling extremely small samples
  • 1 year warranty
More About this Item
Keep a set of these micro spatulas by your balances and microscopes! Use to dispense expensive chemicals, place particulate matter in weighing dishes, apply micro amounts of adhesive for tacking down specimens, or mix small granular samples. Micro spatulas feature a color-coded handle and flatended stainless steel blade. Overall length is 21⁄2" (6.4 cm). Set contains four micro spatulas with 0.1 mm, 0.3 mm, 0.5 mm, and 1.0 mm blade widths.