Cole-Parmer Magnetic Stir Bar, Double Ended, 8 mm x 55 mm

Representative image only.
Cole-Parmer Item # WW-06061-82
Barbell shape stirs even your thickest suspensions

SPECS & DESCRIPTION

Specifications & Description

  • Length (in)5/16
  • Length (cm)0.8
  • Diameter (in)2.165
  • Diameter (cm)5.5
  • DescriptionMagnetic Stir Bar, Double Ended

More About this Item

The double-ended PTFE coated magnetic stir bar is ideal for mixing thick heterogeneous mixtures or viscous liquids for various applications including chemical and biological experiments, buffer solutions, and bacterial growth media. It is designed to stir over a long period of time, is easy to clean, and offers minimal risk of contaminating the solution.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat