Cole-Parmer Loop-Powered Panel Mount Process Meter, NEMA 4X with backlight and heater

Representative image only.
Cole-Parmer Item # WW-30007-54
  • 3 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • Control Input mA4-20
  • Display Type0.6\" LCD
  • Display Digits3.5
  • Max Operating Temperature (° F)185
  • Min Operating Temperature (° F)-67
  • EnclosureNEMA 4; IP66
  • Overall Width (in)3 5/32
  • Overall Height (in)4 21/64
  • Overall Depth (in)2 49/64
  • Overall Width (mm)80
  • Overall Height (mm)110
  • Overall Depth (mm)70.1
  • DescriptionLoop-Powered Panel Mount Process Meter, NEMA 4X with backlight and heater
  • Warranty3 Years
3 Year WarrantyConformite EuropeenneIP 65

Key Features

  • 3 year warranty

More About this Item

All power comes directly from the 4 to 20 mA input signal—no additional AC power source is required! Explosion-proof models provide safe use in hazardous locations and are easy to install and calibrate. These rugged, yet economical meters are ideal for use in hostile environments. Choose the backlit display option when mounting the meters in poor lighting conditions. Meters and backlighting option are powered directly from the 4 to 20 mA loop; no need for an AC power source, batteries, or additional wiring. Models with explosion-proof enclosures meet specifications for use in Class I, groups B, C, D and Class II, for Groups E, F, and G locations.

These 3-1⁄2 digit loop-powered meters are perfect for applications where a simple, economical display is required and AC power is not available. Meters are housed in a rugged NEMA 4X (IP66) enclosure and have one conduit hole. Choose a model with loop-powered backlighting to easily see the display in any lighting condition. The optional 24 VDC powered heater allows the meter to operate in temperatures as low as —67°F —55°C).

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat