Cole-Parmer Liquid Sampler Kit; HDPE
Representative image only.
Cole-Parmer Item # WW-66501-15
Versatile sampling kits with interchangeable parts
  • All components made of FDA-compliant HDPE
  • Purchase individual replacement components as needed!
SPECS & DESCRIPTION
Specifications & Description
  • MaterialHDPE
  • SterileNo
  • DescriptionLiquid Sampler Kit; HDPE
Key Features
  • All components made of FDA-compliant HDPE
  • Purchase individual replacement components as needed!
More About this Item
All dipping beakers, stir blades, and sampling bottles use the same thread to be compatible with all three handle length options. Replacement handles and samplers may be replaced as needed — no need to purchase an entire kit if a part is lost or worn!
Liquid Sampler Kit includes:
One 1.5 ft (0.45 m) handle
One 3 ft (0.9 m) handle
One 6 ft (1.8 m) handle
One liquid sampler adapter with 100 mL bottle and cap
One liquid sampler adapter with 500 mL bottle and cap
The liquid sampler adapters connect the handles to the bottles, and have four small holes to collect liquid samples in the bottles. After capture, store your sample for further analysis with the included bottle cap.