Cole-Parmer Vial Limited-Volume Insert, Bottom Spring, Glass, 250 µL; 100/Pk
Representative image only.
Cole-Parmer Item # WW-98730-00
Keep your samples safe with these vial accessories
  • Select from a variety of caps, septa and inserts that fit your application and needs.
  • This includes the low-volume insert only—vial and closure sold separately
  • Limited Lifetime Warranty
PRODUCT OPTIONS
₹13,300.00INR / Pkg of 100
SPECS & DESCRIPTION
Specifications & Description
  • Volume (mL)0.25
  • Cap TypeLimited-Volume Insert
  • Cap colorClear
  • DescriptionLimited-Volume Insert, Glass w/Bottom Spring, 250 µL
  • WarrantyLimited Lifetime
Limited Lifetime Warranty
Key Features
  • Select from a variety of caps, septa and inserts that fit your application and needs.
  • This includes the low-volume insert only—vial and closure sold separately
  • Limited Lifetime Warranty
More About this Item
Suitable for use with Cole-Parmer® Versi-Vials
Inserts come preassembled with a polymer bottom spring. The spring acts as a shock absorber that protects against breakage if the needle bottoms out. Conical design permits complete sample removal.