Cole-Parmer In-Line pH Electrode with Tuff-Tip, No ATC Element, tinned ends
Representative image only.
Cole-Parmer Item # WW-05995-40
Semi-domed, tuff-tip limits glass exposure and protects electrode from damage
  • Order single or double junction disposable electrodes for your in-line process control needs
  • Available with BNC or tinned ends
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Electrode TypeSingle junction
  • Electrode Body MaterialPVDF
  • Min pH (pH)0
  • Max pH (pH)13
  • Min Temperature (° C)-5
  • Max Temperature (° C)80
  • GuardTuff-Tip
  • Thread Size NPT(M) (in)1/2
  • ConnectorTinned ends
  • Cable Length (m)3
  • DescriptionIn-Line pH Electrode with Tuff-Tip, No ATC Element, tinned ends
  • Warranty1 year
1 Year Warranty
Key Features
  • Order single or double junction disposable electrodes for your in-line process control needs
  • Available with BNC or tinned ends
  • 1 year warranty
More About this Item
Electrodes feature PVDF housing with 1/2'' NPT(M) threads and withstands temperatures from 23 to 176°F (--5 to 80°C) and pressures to 100 psi. Electrodes include 10-ft cable with BNC pH connector and tinned ends for ATC element. Insertion depth 1''.