Cole-Parmer High-Throughput Horizontal Gel System

Representative image only.
Cole-Parmer Item # WW-28562-00
For the rapid screening of samples loaded directly from microcell plates
  • UV-transparent acrylic gel tray has “anchors” to prevent gel from floating
  • 3 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • Buffer Solution Required (mL)3000
  • Gel Length (cm)26
  • Gel Width (cm)26
  • Cell Width (in)18.88
  • Cell Depth (in)11
  • Cell Width (cm)14.96
  • Cell Height (cm)10.16
  • Cell Depth (cm)27.94
  • DescriptionHigh-Throughput Horizontal Gel System
  • Warranty3 Years
3 Year Warranty

Key Features

  • UV-transparent acrylic gel tray has “anchors” to prevent gel from floating
  • 3 year warranty

More About this Item

Multiple comb slots let you configure gels for several simultaneous short runs—ideal for screening large numbers of samples. A separate cooling chamber allows flow of water underneath gel bed to maintain constant temperature. The tapered-baffle design lets you cast gel directly in the unit.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat